Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENO3 Monoclonal Antibody (02)
GREENER_CHOICE

Catalog No. PIMA529238
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA529238 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA529238 Supplier Invitrogen™ Supplier No. MA529238
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ENO3/beta-enolase.

This gene encodes one of the three enolase isoenzymes found in mammals. This isoenzyme is found in skeletal muscle cells in the adult where it may play a role in muscle development and regeneration. A switch from alpha enolase to beta enolase occurs in muscle tissue during development in rodents. Mutations in this gene have be associated glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENO3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 02
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ENO3
Gene Accession No. P13929
Gene Alias 2-phospho-D-glycerate hydrolyase; 2-phospho-D-glycerate hydro-lyase; BBE; beta beta enolase; beta-enolase; cb883; ENO1; ENO3; Eno-3; eno3 protein; enob; enolase 1, (alpha); enolase 3; enolase 3 (beta, muscle); enolase 3, (beta, muscle); enolase 3, beta muscle; enolase 3, beta, muscle; fj24f12; GSD13; hypothetical protein LOC540303; MSE; muscle enriched enolase; Muscle specific enolase (beta beta enolose) (two splicing products); muscle-specific enolase; Skeletal muscle enolase; wu:fj24f12
Gene Symbols ENO3
Host Species Mouse
Immunogen Recombinant Human ENO3/beta-enolase protein (Met1-Lys434).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2027
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.