Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENO3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120444
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120444 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120444 Supplier Invitrogen™ Supplier No. PA5120444
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse heart, Mouse skeletal muscle, Rat heart, Rat skeletal muscle. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: MAMQKIFARE ILDSRGNPTV EVDLHTAKGR FRAAVPSGAS TGIYEALELR DGDKGRYLGK GVLKAVENIN NTLGPALLQK KLSVVDQEKV DKFMIELDGT ENKSKFGANA ILGVSLAVCK AGAAEKGVPL YRHIADLAGN PDLILPVPAF NVINGGSHAG NKLAMQEFMI LPVGASSFKE AMRIGAEVYH.

This gene encodes one of the three enolase isoenzymes found in mammals. This isoenzyme is found in skeletal muscle cells in the adult where it may play a role in muscle development and regeneration. A switch from alpha enolase to beta enolase occurs in muscle tissue during development in rodents. Mutations in this gene have be associated glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENO3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ENO3
Gene Accession No. P13929, P15429, P21550
Gene Alias 2-phospho-D-glycerate hydrolyase; 2-phospho-D-glycerate hydro-lyase; BBE; beta beta enolase; beta-enolase; cb883; ENO1; ENO3; Eno-3; eno3 protein; enob; enolase 1, (alpha); enolase 3; enolase 3 (beta, muscle); enolase 3, (beta, muscle); enolase 3, beta muscle; enolase 3, beta, muscle; fj24f12; GSD13; hypothetical protein LOC540303; MSE; muscle enriched enolase; Muscle specific enolase (beta beta enolose) (two splicing products); muscle-specific enolase; Skeletal muscle enolase; wu:fj24f12
Gene Symbols ENO3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-190 of human ENO3 (NP_001967.3).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13808, 2027, 25438
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.