Learn More
Invitrogen™ ENO3 Polyclonal Antibody

Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (97%), Rat (97%), Zebrafish (85%), Xenopus laevis (89%), Pig (97%), Rabbit (97%), Chicken (82%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | ENO3 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | ENO3 |
| Gene Accession No. | P13929 |
| Gene Alias | 2-phospho-D-glycerate hydrolyase; 2-phospho-D-glycerate hydro-lyase; BBE; beta beta enolase; beta-enolase; cb883; ENO1; ENO3; Eno-3; eno3 protein; enob; enolase 1, (alpha); enolase 3; enolase 3 (beta, muscle); enolase 3, (beta, muscle); enolase 3, beta muscle; enolase 3, beta, muscle; fj24f12; GSD13; hypothetical protein LOC540303; MSE; muscle enriched enolase; Muscle specific enolase (beta beta enolose) (two splicing products); muscle-specific enolase; Skeletal muscle enolase; wu:fj24f12 |
| Gene Symbols | ENO3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.