Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENO3 Antibody (008), Alexa Fluor™ 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290275AF594
Description
ENO3 Monoclonal specifically detects ENO3 in Human samples. It is validated for ELISA.Specifications
ENO3 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
8 | |
ELISA | |
2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, beta-enolase, EC 4.2.1, EC 4.2.1.11, Enolase 3, enolase 3 (beta, muscle), enolase 3, (beta, muscle), GSD13, MSE, Muscle-specific enolase, Skeletal muscle enolase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ENO3 (AAH17249.1; Met1-Lys434). | |
0.1 mL | |
metabolism | |
2027 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction