Learn More
Invitrogen™ IL-9 (Enokizumab Biosimilar) Recombinant Human Monoclonal Antibody

Description
Endotoxin level <0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt. This product is a Humanized mouse antibody.
Specifications
Specifications
| Antigen | IL-9 (Enokizumab Biosimilar) |
| Applications | ELISA, SDS-Page |
| Classification | Recombinant Monoclonal |
| Concentration | 1.52 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Il9 |
| Gene Accession No. | P15248 |
| Gene Alias | cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40 |
| Gene Symbols | Il9 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.