Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Enolase 1 Antibody (1G7), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259457C
Description
Enolase 1 Monoclonal antibody specifically detects Enolase 1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
| Enolase 1 | |
| Monoclonal | |
| DyLight 650 | |
| 2-phospho-D-glycerate hydro-lyase, Alpha enolase, C-myc promoter-binding protein, c-myc promoter-binding protein-1, EC 4.2.1, EC 4.2.1.11, ENO1L1, Enolase 1, enolase 1, (alpha), MBP-1, MBPB1, MPB-1, MPB1c-myc promoter-binding protein 1, MYC promoter-binding protein 1, NNE, Non-neural enolase, Phosphopyruvate hydratase, Plasminogen-binding protein, PPHalpha enolase like 1, tau-crystallin | |
| Recombinant human Enolase 1 (1-434aa) purified from E. coli | |
| 0.1 mL | |
| Cancer, Core ESC Like Genes, Stem Cell Markers | |
| 2023 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 1G7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction