Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Enolase 1 Antibody (1G7), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259457PE
This item is not returnable.
View return policy
Description
Enolase 1 Monoclonal antibody specifically detects Enolase 1 in Human samples. It is validated for Flow CytometrySpecifications
Enolase 1 | |
Monoclonal | |
PE | |
2-phospho-D-glycerate hydro-lyase, Alpha enolase, C-myc promoter-binding protein, c-myc promoter-binding protein-1, EC 4.2.1, EC 4.2.1.11, ENO1L1, Enolase 1, enolase 1, (alpha), MBP-1, MBPB1, MPB-1, MPB1c-myc promoter-binding protein 1, MYC promoter-binding protein 1, NNE, Non-neural enolase, Phosphopyruvate hydratase, Plasminogen-binding protein, PPHalpha enolase like 1, tau-crystallin | |
Recombinant human Enolase 1 (1-434aa) purified from E. coli | |
0.1 mL | |
Cancer, Core ESC Like Genes, Stem Cell Markers | |
2023 | |
Store at 4°C in the dark. | |
IgG2a κ |
Flow Cytometry | |
1G7 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction