Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Enolase 1 Antibody (253), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP225147AF488
Description
Enolase 1 Monoclonal specifically detects Enolase 1 in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Enolase 1 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ENO1 | |
The N-terminal 12 amino acids of bovine Enolase 1, which was synthesized on a 8-amine lysine core. [UniProt# Q9XSJ4] | |
0.1 mL | |
Cancer, Core ESC Like Genes, Stem Cell Markers | |
2023 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
253 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
2-phospho-D-glycerate hydro-lyase, Alpha enolase, C-myc promoter-binding protein, c-myc promoter-binding protein-1, EC 4.2.1, EC 4.2.1.11, ENO1L1, Enolase 1, enolase 1, (alpha), MBP-1, MBPB1, MPB-1, MPB1c-myc promoter-binding protein 1, MYC promoter-binding protein 1, NNE, Non-neural enolase, Phosphopyruvate hydratase, Plasminogen-binding protein, PPHalpha enolase like 1, tau-crystallin | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Bovine, Equine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction