Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enolase 2/Neuron-specific Enolase Antibody (ENO2/1375), Novus Biologicals™
SDP

Catalog No. p-7105629 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247635F 0.02 mg
NBP247635A 0.1 mg
NBP247635B 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP247635F Supplier Novus Biologicals Supplier No. NBP2476350.02MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Enolase 2/Neuron-specific Enolase Monoclonal specifically detects Enolase 2/Neuron-specific Enolase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).

Specifications

Antigen Enolase 2/Neuron-specific Enolase
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence
Classification Monoclonal
Clone ENO2/1375
Conjugate Unconjugated
Dilution Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry-Paraffin 0.5-1ug/ml, SDS-Page, Immunofluorescence 1-2ug/ml
Formulation 10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias 2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE
Gene Symbols ENO2
Host Species Mouse
Immunogen A synthetic peptide corresponding to aa416-433 of human NSE gamma.
Molecular Weight of Antigen 50 kDa
Purification Method Protein A or G purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2026
Test Specificity Recognizes a protein of about 50kDa, which is identified as gamma-enolase. Three isoenzymes of enolases are identified, alpha, beta and gamma. Alpha-isoform is expressed in most tissues, whereas beta-form is expressed predominantly in muscle tissue whereas gamma-enolase is found only in nervous tissue. These isoforms exist as both homodimers and heterodimers, and they play a role in converting phosphoglyceric acid to phosphenolpyruvic acid in the glycolytic pathway. NSE-gamma is a useful marker to identify peripheral nerves and tumors of neuro-endocrine origins, such as pheochromocytomas. It it be usually employed in combination with other markers such as Synaptophysin, Chromogranin A, and Neurofilament.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.