Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Enolase 2/Neuron-specific Enolase Antibody (NSEP1), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250532MFV500
Description
Enolase 2/Neuron-specific Enolase Monoclonal antibody specifically detects Enolase 2/Neuron-specific Enolase in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| Enolase 2/Neuron-specific Enolase | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| NSEP1 | |
| Western Blot, ELISA, Immunohistochemistry | |
| 2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE | |
| Ovalbumin-conjugated synthetic peptides corresponding to human NSE amino acid sequence:NSE-P1: aa's 416-433 - LGDEARFAGHNFRNPSVL | |
| 0.1 mL | |
| Cancer, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
| 2026 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction