Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Enolase 2/Neuron-specific Enolase Antibody (NSEP2), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250533AF488
Description
Enolase 2/Neuron-specific Enolase Monoclonal specifically detects Enolase 2/Neuron-specific Enolase in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Enolase 2/Neuron-specific Enolase | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ENO2 | |
Ovalbumin-conjugated synthetic peptides corresponding to human NSE amino acid sequence:NSE-P2: aa's 271-285 - TGDQLGALYQDFVRD | |
0.1 mL | |
Cancer, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
2026 | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
NSEP2 | |
Western Blot, ELISA, Immunohistochemistry | |
2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Not reactive with -isozyme of enolase. | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction