Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENOPH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100501
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100501 Supplier Invitrogen™ Supplier No. PA5100501
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ENOPH1.

Bifunctional enzyme that catalyzes the enolization of 2,3-diketo-5-methylthiopentyl-1-phosphate (DK-MTP-1-P) into the intermediate 2-hydroxy-3-keto-5-methylthiopentenyl-1-phosphate (HK-MTPenyl-1-P), which is then dephosphorylated to form the acireductone 1,2-dihydroxy-3-keto-5-methylthiopentene (DHK-MTPene).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENOPH1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ENOPH1
Gene Accession No. Q5PPH0, Q8BGB7, Q9UHY7
Gene Alias 2,3-diketo-5-methylthio-1-phosphopentane phosphatase; 2,3-diketo-5-methylthio-1-phosphopentane phosphatase {ECO:0000255; 2310057D15Rik; acireductone synthase; BB183658; C81437; E1; E-1 enzyme; enolase-phosphatase 1; enolase-phosphatase E1; enolase-phosphatase E1 {ECO:0000255; ENOPH1; HAMAP-Rule:MF_03117}; Masa; MASA homolog; MASA homolog {ECO:0000255; MST145; MSTP145; mtnC; RGD1309016
Gene Symbols ENOPH1
Host Species Rabbit
Immunogen A synthesized peptide derived from human ENOPH1(Accession Q9UHY7), corresponding to amino acid residues T211-T261.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 305177, 58478, 67870
Target Species Human, Mouse, Monkey, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.