Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eNOS Antibody (6H2), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP151582AF405
Description
eNOS Monoclonal specifically detects eNOS in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
eNOS | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NOS3 | |
Purified recombinant fragment of human eNOS expressed in E. coli. [UniProt# P29474] | |
0.1 mL | |
Cancer, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific | |
4846 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
6H2 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOSIII | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction