Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eNOS Antibody (6H2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$181.72 - $492.89
Specifications
| Antigen | eNOS |
|---|---|
| Clone | 6H2 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2 ug/ml, ELISA 1:10000, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP151582SS
![]() |
Novus Biologicals
NBP151582SS |
0.025 mL |
Each for $181.72
|
|
|||||
NBP151582
![]() |
Novus Biologicals
NBP151582 |
0.1 mL |
Each for $492.89
|
|
|||||
Description
eNOS Monoclonal specifically detects eNOS in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| eNOS | |
| 1.0 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOSIII | |
| NOS3 | |
| IgG1 | |
| Ammonium sulfate precipitation | |
| 133 kDa |
| 6H2 | |
| Western Blot 2 ug/ml, ELISA 1:10000, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 | |
| Monoclonal | |
| Precipitation | |
| RUO | |
| P29474 | |
| 4846 | |
| Purified recombinant fragment of human eNOS expressed in E. coli. [UniProt# P29474] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title