Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eNOS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA516887
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA516887 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA516887 Supplier Invitrogen™ Supplier No. PA516887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA5-16887 targets eNOS in immunohistochemistry (paraffin) applications and shows reactivity with Bovine, Canine, Human, mouse, Porcine, and Rat samples. Suggested positive control is capillary endothelium and regenerative endothelium. The PA5-16887 immunogen is a synthetic peptide derived from C-ternimal of human eNOS.

Nitric oxide (NO) is an inorganic, gaseous free radical that carries a variety of messages between cells. Vasorelaxation, neurotransmission and cytotoxicity can all be potentiated through cellular response to NO. NO production is mediated by members of the nitric oxide synthase (NOS) family. NOS catalyzes the oxidization of L-arginine to produce L-citrulline and NO. Two constitutive isoforms, brain or neuronal NOS (b or nNOS, type I) and endothelial cell NOS (eNOS, type III), and one inducible isoform (iNOS, type II), have been cloned. All NOS isoforms contain calmodulin, nicotinamide adenine dinucleotide phosphate (NADPH), flavin adenine dinucleotide (FAD), and flavin mononucleotide (FMN) binding domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eNOS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.081 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.6
Gene NOS3
Gene Accession No. P29474
Gene Alias 2310065A03Rik; cNOS; Constitutive NOS; Ecnos; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; endothelial nitric oxide synthase NOS3; endothelial NOS; eNos; e-NOS; nitric oxide synthase; nitric oxide synthase 3; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; nitric oxide synthase 3, endothelial cell; nitric oxide synthase, endothelial; NOS; NOS type III; Nos3; Nos-3; NOSIII; OTTHUMP00000213183
Gene Symbols NOS3
Host Species Rabbit
Immunogen A synthetic peptide derived from C-ternimal of human eNOS.
Purification Method Affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4846
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.