Learn More
Invitrogen™ ENOSF1 Polyclonal Antibody

Description
Immunogen sequence: QHLIIFDYIS VSASLENRVC EYVDHLHEHF KYPVMIQRAS YMPPKDPGYS TEMKEESVKK HQYPDGEVWK K Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 25%.
Specifications
Specifications
| Antigen | ENOSF1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ENOSF1 |
| Gene Accession No. | Q7L5Y1 |
| Gene Alias | antisense RNA to thymidylate synthase; enolase superfamily member 1; ENOSF1; HSRTSBETA; L-fuconate dehydratase; mitochondrial enolase superfamily member 1; rTS; rTS alpha; rTS beta; TYMSAS |
| Gene Symbols | ENOSF1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.