Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ENOSF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599877
Description
Antibody detects endogenous levels of total ENOSF1.
DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. Human proteins MECP2, MBD1, MBD2, MBD3, and MBD4 comprise a family of nuclear proteins related by the presence in each of a methyl-CpG binding domain (MBD). Each of these proteins, with the exception of MBD3, is capable of binding specifically to methylated DNA. MECP2, MBD1 and MBD2 can also repress transcription from methylated gene promoters. In contrast to other MBD family members, MECP2 is X-linked and subject to X inactivation. MECP2 is dispensible in stem cells, but is essential for embryonic development. MECP2 gene mutations are the cause of most cases of Rett syndrome, a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females.
Specifications
ENOSF1 | |
Polyclonal | |
Unconjugated | |
ENOSF1 | |
antisense RNA to thymidylate synthase; enolase superfamily member 1; ENOSF1; HSRTSBETA; L-fuconate dehydratase; mitochondrial enolase superfamily member 1; rTS; rTS alpha; rTS beta; TYMSAS | |
Rabbit | |
Affinity chromatography | |
RUO | |
55556 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q7L5Y1 | |
ENOSF1 | |
A synthesized peptide derived from human ENOSF1(Accession Q7L5Y1), corresponding to amino acid residues V388-L438. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction