Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Invitrogen™ ENOX1 Polyclonal Antibody
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597708
Description
ENOX1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Probably acts as a terminal oxidase of plasma electron transport from cytosolic NAD(P)H via hydroquinones to acceptors at the cell surface. Hydroquinone oxidase activity alternates with a protein disulfide-thiol interchange/oxidoreductase activity which may control physical membrane displacements associated with vesicle budding or cell enlargement. The activities oscillate with a period length of 24 minutes and play a role in control of the ultradian cellular biological clock.
            Specifications
| ENOX1 | |
| Polyclonal | |
| Unconjugated | |
| ENOX1 | |
| bA64J21.1; candidate growth-related and time keeping constitutive hydroquinone (NADH) oxidase; candidate growth-related and time keeping constitutive hydroquinone [NADH] oxidase; cCNOX; Cell proliferation-inducing gene 38 protein; CNOX; Constitutive Ecto-NOX; ecto-NOX disulfide-thiol exchanger 1; ENOX1; Hydroquinone [NADH] oxidase; PIG38; Protein disulfide-thiol oxidoreductase | |
| Rabbit | |
| Protein G | |
| RUO | |
| 55068 | |
| -20°C or -80°C if preferred | |
| Liquid | 
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8TC92 | |
| ENOX1 | |
| Recombinant Human Ecto-NOX disulfide-thiol exchanger 1 protein (101-340AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG | 
Safety and Handling
                                                    
                                                    WARNING: Cancer - www.P65Warnings.ca.gov 
                                                
                                            
                                        
                                    
                                
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction