Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENOX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545041
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545041 Supplier Invitrogen™ Supplier No. PA545041
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: YRIRLGSSTD KKDTGRLHVD FAQARDDLYE WECKQRMLAR EERHRRRMEE Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

This gene is a tumor-specific member of the ECTO-NOX family of genes that encode cell surface NADH oxidases. The encoded protein has two enzymatic activities: catalysis of hydroquinone or NADH oxidation, and protein disulfide interchange. The protein also displays prion-like properties. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENOX2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Enox2
Gene Accession No. Q16206
Gene Alias APK1; APK1 antigen; Cova1; cytosolic ovarian carcinoma antigen 1; Ecto-NOX disulfide-thiol exchanger 2; enox2; Hydroquinone [NADH] oxidase; Protein disulfide-thiol oxidoreductase; RGD1563643; tNOX; tumor-associated hydroquinone oxidase; zmp:0000001197
Gene Symbols Enox2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ENOX2 (aa 184-233).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10495
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.