Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEP ENPAC™ Spillpal Drum Pallets
SDP

Catalog No. p-3557048
Encompass
Click to view available options
Dimensions (L x W x H):
2 x 2 x 4 in.
2 x 4 x 4 in.
2 x 8 x 4 in.
4 x 4 x 4 in.
4 x 6 x 4 in.
4 x 8 x 4 in.
Material:
Nylon
Polyethylene
Type:
1-drum Spill Pallet
2-drum Spill Pallet
4-drum In-line Spill Pallet
4-drum Spill Pallet
6-drum Spill Pallet
8-drum Spill Pallet

Provides incidental spill control during temporary storage of drums, machine parts, tanks and generators

  • Flexible four inch foam sidewalls spring back to form, allowing easy drive-in and drive-out with a hand truck or forklift
  • Designed for 37.8L (10 gal.) drums
  • Able to be folded and stored
  • Reusable

Specifications

Certifications/Compliance SPCC, EPA, NPDES
Color Yellow
Capacity (English) 40 gal.
Capacity (Metric) 113.5L
Material Nylon
Type 4-drum In-line Spill Pallet
For Use With (Application) Used to store hazardous material
Holds 4 Drums
Includes Foam sidewalls
Dimensions (L x W x H) 2 x 8 x 4 in.
Length (English) 2 ft.
Length (Metric) 0.6 m
Width (English) 8 ft.
Width (Metric) 2.43 m
Height (English) 4 in.
Height (Metric) 10.16 cm
Weight (English) 5.5 lb.
Weight (Metric) 2.49 kg
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.