Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENPEP Monoclonal Antibody (OTI1D4), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA504037

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-4810


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

CD249 appears to have a role in the catabolic pathway of the renin-angiotensin system. Probably plays a role in regulating growth and differentiation of early B-lineage cells.
Specifications

Specifications

ENPEP
Monoclonal
0.88 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P16406, Q07075
ENPEP
Full length human recombit protein of human ENPEP produced in HEK293T cell.
100 μL
Primary
Human, Mouse
Antibody
IgG2a
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1D4
Unconjugated
ENPEP
APA, CD249, gp160
Mouse
Affinity Chromatography
RUO
13809, 2028
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.