Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENPP-1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB600816AF750
Description
ENPP-1 Polyclonal specifically detects ENPP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
ENPP-1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA | |
ectonucleotide pyrophosphatase/phosphodiesterase 1, E-NPP 1, Ly-41 antigen, M6S1NPP1, Membrane component chromosome 6 surface marker 1, membrane component, chromosome 6, surface marker 1, NPPSalkaline phosphodiesterase 1, PC1, PC-1, PCA1ARHR2, PDNP1ectonucleotide pyrophosphatase/phosphodiesterase family member 1, Phosphodiesterase I/nucleotide pyrophosphatase 1, plasma-cell membrane glycoprotein 1, Plasma-cell membrane glycoprotein PC-1 | |
Goat | |
0.1 mL | |
5167 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
ENPP1 | |
A synthetic peptide made to a C-terminal portion of the human ENPP1 protein (between residues 900-925) [UniProt P22413] | |
Primary | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction