Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENPP-2/Autotaxin Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298332F
Description
ENPP-2/Autotaxin Polyclonal specifically detects ENPP-2/Autotaxin in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ENPP-2/Autotaxin | |
Polyclonal | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
ATXFLJ26803, ATX-X, AUTOTAXIN, autotaxin-t, EC 3.1.4.39, ectonucleotide pyrophosphatase/phosphodiesterase 2, ectonucleotide pyrophosphatase/phosphodiesterase family member 2, E-NPP 2, Extracellular lysophospholipase D, LysoPLD, PD-IALPHA, PDNP2NPP2, phosphodiesterase I/nucleotide pyrophosphatase 2, plasma lysophospholipase D | |
Produced in rabbits immunized with purified, recombinant ENPP-2/Autotaxin (Q9R1E6-1; Ser49-Ile862). | |
0.1 mL | |
Cardiovascular Biology | |
5168 | |
Store at 4°C in the dark. | |
IgG |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
FITC | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction