Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENPP-3/CD203c Antibody (3B2), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | ENPP-3/CD203c |
---|---|
Clone | 3B2 |
Dilution | Flow Cytometry, ELISA |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
ENPP-3/CD203c Monoclonal antibody specifically detects ENPP-3/CD203c in Human, Cynomolgus Monkey samples. It is validated for Flow Cytometry, ELISASpecifications
ENPP-3/CD203c | |
Flow Cytometry, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human, Cynomolgus Monkey | |
CD203c, CD203c antigen, dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3), dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3), ectonucleotide pyrophosphatase/phosphodiesterase 3, ectonucleotide pyrophosphatase/phosphodiesterase family member 3, E-NPP 3, gp130RB13-6, NPP3, PD-IBETA, PDNP3B10, Phosphodiesterase I beta, Phosphodiesterase I/nucleotide pyrophosphatase 3, phosphodiesterase-I beta | |
Recombinant Human ENPP-3/CD203c protein [UniProt O14638] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
3B2 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Human | |
Protein Phosphatase | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
5169 | |
IgG1 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title