Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENPP-3/CD203c Antibody (NP4D6), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP144643AF700
Description
ENPP-3/CD203c Monoclonal specifically detects ENPP-3/CD203c in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
ENPP-3/CD203c | |
Monoclonal | |
Alexa Fluor 700 | |
CD203c, CD203c antigen, dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3), dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3), ectonucleotide pyrophosphatase/phosphodiesterase 3, ectonucleotide pyrophosphatase/phosphodiesterase family member 3, E-NPP 3, gp130RB13-6, NPP3, PD-IBETA, PDNP3B10, Phosphodiesterase I beta, Phosphodiesterase I/nucleotide pyrophosphatase 3, phosphodiesterase-I beta | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
The mouse monoclonal antibody NP4D6 reacts with CD203c, a transmembrane ectoenzyme expressed on basophils and mast cells, and overexpressed upon their activation. HLDA VIII | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
NP4D6 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
ENPP3 | |
HEK-293 cells transfected with human CD203c | |
0.1 mL | |
Protein Phosphatase | |
5169 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction