Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENPP-5 Antibody (014), FITC , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289909F
Description
ENPP-5 Monoclonal antibody specifically detects ENPP-5 in Human samples. It is validated for ELISA, ELISASpecifications
ENPP-5 | |
Monoclonal | |
FITC | |
EC 3.1, ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative function), ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative), ectonucleotide pyrophosphatase/phosphodiesterase family member 5, E-NPP 5, KIAA0879, NPP-5 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human ENPP-5 (Q9UJA9; Met1-Gly429). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
014 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
59084 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction