Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENPP-5 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299809F
Description
ENPP-5 Polyclonal specifically detects ENPP-5 in Human samples. It is validated for ELISA.Specifications
ENPP-5 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
59084 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
FITC | |
EC 3.1, ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative function), ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative), ectonucleotide pyrophosphatase/phosphodiesterase family member 5, E-NPP 5, KIAA0879, NPP-5 | |
Produced in rabbits immunized with purified, recombinant ENPP-5 (Q9UJA9; Met1-Gly429). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction