Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENPP-5 Rabbit anti-Human, Clone: 14, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289909100UL
Description
ENPP-5 Monoclonal specifically detects ENPP-5 in Human samples. It is validated for Sandwich ELISA Capture.Specifications
ENPP-5 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
59084 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
ELISA | |
14 | |
Sandwich ELISA Capture 1:250-1:2000 | |
EC 3.1, ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative function), ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative), ectonucleotide pyrophosphatase/phosphodiesterase family member 5, E-NPP 5, KIAA0879, NPP-5 | |
Recombinant Human ENPP-5 Protein | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction