Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENPP-7/Alk-SMase Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299660MFV610
Description
ENPP-7/Alk-SMase Polyclonal antibody specifically detects ENPP-7/Alk-SMase in Human samples. It is validated for Western Blot, ELISASpecifications
ENPP-7/Alk-SMase | |
Polyclonal | |
Western Blot, ELISA | |
Alkaline sphingomyelin phosphodiesterase, alkaline sphingomyelinase, ALK-SMase, EC 3.1.4.12, ectonucleotide pyrophosphatase/phosphodiesterase 7, ectonucleotide pyrophosphatase/phosphodiesterase family member 7, E-NPP 7, Intestinal alkaline sphingomyelinase, MGC50179, NPP-7 | |
Produced in rabbits immunized with purified, recombinant Human ENPP-7/Alk-SMase (Accession#: NP_848638.2; Met 1-Ser 439) | |
0.1 mL | |
Lipid and Metabolism | |
339221 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction