Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENPP-7/Alk-SMase Rabbit anti-Human, Clone: 1, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$353.50 - $560.50

Specifications

Antigen ENPP-7/Alk-SMase
Clone 1
Dilution ELISA 1:5000-1:10000
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB020872
SDP
View Documents
Novus Biologicals
NBP28969450UL
50 μg
Each for $353.50
Only null left
Add to Cart
 
NB020871
SDP
View Documents
Novus Biologicals
NBP289694100UL
100 μg
Each for $560.50
Only null left
Add to Cart
 
Description

Description

ENPP-7/Alk-SMase Monoclonal specifically detects ENPP-7/Alk-SMase in Human samples. It is validated for ELISA.
Specifications

Specifications

ENPP-7/Alk-SMase
ELISA 1:5000-1:10000
Monoclonal
Purified
RUO
Human
Alkaline sphingomyelin phosphodiesterase, alkaline sphingomyelinase, ALK-SMase, EC 3.1.4.12, ectonucleotide pyrophosphatase/phosphodiesterase 7, ectonucleotide pyrophosphatase/phosphodiesterase family member 7, E-NPP 7, Intestinal alkaline sphingomyelinase, MGC50179, NPP-7
Recombinant Human ENPP-7/Alk-SMase protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
1
ELISA
Unconjugated
Rabbit
Lipid and Metabolism
0.2 um filtered solution in PBS
339221
IgG
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.