Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENPP1/PC1/3 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS1760R

Description
This gene is a member of the ecto-nucleotide pyrophosphatase/phosphodiesterase (ENPP) family. The encoded protein is a type II transmembrane glycoprotein comprising two identical disulfide-bonded subunits. This protein has broad specificity and cleaves a variety of substrates, including phosphodiester bonds of nucleotides and nucleotide sugars and pyrophosphate bonds of nucleotides and nucleotide sugars. This protein may function to hydrolyze nucleoside 5′ triphosphates to their corresponding monophosphates and may also hydrolyze diadenosine polyphosphates. Mutations in this gene have been associated with ′idiopathic′ infantile arterial calcification, ossification of the posterior longitudinal ligament of the spine (OPLL), and insulin resistance.Specifications
ENPP1/PC1/3 | |
Polyclonal | |
Unconjugated | |
ENPP1 | |
4833416E15Rik; AI428932; alkaline phosphodiesterase 1; Alkaline phosphodiesterase I; ARHR2; C76301; CD203c; COLED; ectonucleotide pyrophosphatase/phosphodiesterase 1; Ectonucleotide pyrophosphatase/phosphodiesterase family member 1; Ectonucleotide pyrophosphatase/phosphodiesterase family member 1, secreted form; E-NPP 1; ENPP1; E-NPP1; HGNC:3356; Ly-41; Ly-41 antigen; Lymphocyte antigen 41; M6S1; Membrane component chromosome 6 surface marker 1; membrane component, chromosome 6, surface marker 1; NPP1; NPPase; Npps; Nucleotide diphosphatase; Nucleotide pyrophosphatase; OTTHUMP00000043194; PC1; PC-1; Pca; PCA1; Pca-1; Pdnp1; Phosphodiesterase I/nucleotide pyrophosphatase 1; plasma-cell membrane glycoprotein 1; Plasma-cell membrane glycoprotein PC-1; ttw; twy | |
Rabbit | |
Protein A | |
RUO | |
5167 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P22413 | |
ENPP1 | |
KLH conjugated synthetic peptide derived from human ENPP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction