Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENPP1 Polyclonal Antibody

Catalog No. PIPA5122014
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122014 20 μL
PIPA5119090 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122014 Supplier Invitrogen™ Supplier No. PA5122014
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ENPP1 Polyclonal Antibody for ICC/IF, IHC (P), Western Blot

This gene is a member of the ecto-nucleotide pyrophosphatase/phosphodiesterase (ENPP) family. The encoded protein is a type II transmembrane glycoprotein comprising two identical disulfide-bonded subunits. This protein has broad specificity and cleaves a variety of substrates, including phosphodiester bonds of nucleotides and nucleotide sugars and pyrophosphate bonds of nucleotides and nucleotide sugars. This protein may function to hydrolyze nucleoside 5' triphosphates to their corresponding monophosphates and may also hydrolyze diadenosine polyphosphates. Mutations in this gene have been associated with 'idiopathic' infantile arterial calcification, ossification of the posterior longitudinal ligament of the spine (OPLL), and insulin resistance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENPP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene ENPP1
Gene Accession No. Q924C3
Gene Alias 4833416E15Rik; AI428932; alkaline phosphodiesterase 1; Alkaline phosphodiesterase I; ARHR2; C76301; CD203c; COLED; ectonucleotide pyrophosphatase/phosphodiesterase 1; ectonucleotide pyrophosphatase/phosphodiesterase family member 1; Ectonucleotide pyrophosphatase/phosphodiesterase family member 1, secreted form; E-NPP 1; Enpp1; E-NPP1; HGNC:3356; Ly-41; Ly-41 antigen; Lymphocyte antigen 41; M6S1; Membrane component chromosome 6 surface marker 1; membrane component, chromosome 6, surface marker 1; NPP1; NPPase; NPPS; Nucleotide diphosphatase; Nucleotide pyrophosphatase; OTTHUMP00000043194; PC1; PC-1; Pca; PCA1; Pca-1; PDNP1; Phosphodiesterase I/nucleotide pyrophosphatase 1; plasma-cell membrane glycoprotein 1; plasma-cell membrane glycoprotein PC-1; ttw; twy
Gene Symbols ENPP1
Host Species Rabbit
Immunogen Recombinant protein Ectonucleotide Pyrophosphatase/Phosphodiesterase 1. The antigen corresponds to amino acid range 568-793 of the target protein.
Purification Method Protein A, Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85496
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.