Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENPP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120818
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120818 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120818 Supplier Invitrogen™ Supplier No. PA5120818
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HepG2, A-549, Mouse kidney. The target is usually found in the following locations: Secreted. Immunogen sequence: DLGCTCDDKV EPKNKLDELN KRLHTKGSTE ERHLLYGRPA VLYRTRYDIL YHTDFESGYS EIFLMPLWTS YTVSKQAEVS SVPDHLTSCV RPDVRVSPSF SQNCLAYKND KQMSYGFLFP PYLSSSPEAK YDAFLVTNMV PMYPAFKRVW NYFQRVLVKK YASERNGVNV ISGPIFDYDY DGLHDTEDKI KQYVEGSSIP VPTHYYSIIT SCLDFTQPAD KCDGPLSVSS FILPHRPDNE ESCNSSEDES KWVEELMKMH TARVRDIEHL TSLDFFRKTS RSYPEILTLK TYLHTYESEI.

The protein encoded by this gene functions as both a phosphodiesterase, which cleaves phosphodiester bonds at the 5' end of oligonucleotides, and a phospholipase, which catalyzes production of lysophosphatidic acid (LPA) in extracellular fluids. LPA evokes growth factor-like responses including stimulation of cell proliferation and chemotaxis. This gene product stimulates the motility of tumor cells and has angiogenic properties, and its expression is upregulated in several kinds of carcinomas. The gene product is secreted and further processed to make the biologically active form. Several alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENPP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ENPP2
Gene Accession No. Q13822, Q9R1E6
Gene Alias ATX; ATX-X; autotaxin; AutotaxinATX; autotaxin-t; ectonucleotide pyrophosphatase/phosphodiesterase 2; ectonucleotide pyrophosphatase/phosphodiesterase family member 2; E-NPP 2; Enpp2; Extracellular lysophospholipase D; FLJ26803; lysoPLD; NPP2; Npps2; PD-IALPHA; PDNP2; phosphodiesterase I/nucleotide pyrophosphatase 2; phosphodiesterase I/nucleotide pyrophosphatase 2 (autotaxin); plasma lysophospholipase D
Gene Symbols ENPP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 564-863 of human ENPP2 (NP_001035181.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18606, 5168
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.