Learn More
Invitrogen™ ENPP4 Polyclonal Antibody

Description
The antibody detects endogenous levels of total ENPP4 protein.
Specifications
Specifications
| Antigen | ENPP4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ENPP4 |
| Gene Accession No. | Q9Y6X5 |
| Gene Alias | 4933413N07Rik; AA986363; AI195364; AP3A hydrolase; AP3Aase; Bis(5'-adenosyl)-triphosphatase enpp4; ectonucleotide pyrophosphatase/phosphodiesterase 4; ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative function); ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative); ectonucleotide pyrophosphatase/phosphodiesterase family member 4; E-NPP 4; ENPP4; E-NPP4; Gm90; KIAA0879; mKIAA0879; NPP4; NPP-4 |
| Gene Symbols | ENPP4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.