Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENPP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553454 Supplier Invitrogen™ Supplier No. PA553454
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DEGWTIVLNE SSQKLGDHGY DNSLPSMHPF LAAHGPAFHK GYKHSTINIV DIYPMMCHIL GLKPHPNNGT FGHTKCLLVD QW Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

NPP4, also known as ENPP4 (ectonucleotide pyrophosphatase/phosphodiesterase family member 4), is a 453 amino acid single-pass type I membrane protein that belongs to the nucleotide pyrophosphatase/phosphodiesterase family. The gene that encodes NPP4 consists of approximately 16, 736 bases and maps to human chromosome 6p21.1. Making up nearly 6% of the human genome, chromosome 6 contains around 1, 200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene, and Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6. Notably, the PARK2 gene, which is associated with Parkinson's disease, and the genes encoding the major histocompatibility complex proteins are also located on chromosome 6. A bipolar disorder susceptibility locus has been identified on the q arm of chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENPP4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ENPP4
Gene Accession No. Q9Y6X5
Gene Alias 4933413N07Rik; AA986363; AI195364; AP3A hydrolase; AP3Aase; Bis(5'-adenosyl)-triphosphatase enpp4; ectonucleotide pyrophosphatase/phosphodiesterase 4; ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative function); ectonucleotide pyrophosphatase/phosphodiesterase 4 (putative); ectonucleotide pyrophosphatase/phosphodiesterase family member 4; E-NPP 4; ENPP4; E-NPP4; Gm90; KIAA0879; mKIAA0879; NPP4; NPP-4
Gene Symbols ENPP4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ENPP4. Recombinant protein control fragment (Product #RP-90624).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22875
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.