Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENPP5 Monoclonal Antibody (1C5-1D7)
GREENER_CHOICE

Catalog No. PIMA522431
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522431 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522431 Supplier Invitrogen™ Supplier No. MA522431
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: QPDQQKVLLV SFDGFRWDYL YKVPTPHFHY IMKYGVHVKQ VTNVFITKTY PNHYTLVTGL FAENHGIVAN DMFDPIRNKS FSLDHMNIYD SKFWEEATPI WITNQRAGHT SGAAMWPGTD VKIHKRFPTH YMPYNESVSF EDRVAKIIEW FTSKEPINLG LLYWEDPDDM GHHLGPDSPL MGPVISDIDK KLGYLIQMLK KAKLWNTLNL IITSDHGMTQ CSEERLIELD QYLDKDHYTL IDQSPVAAIL PKEGKFDEVY EALTHAHPNL TVYKKEDVPE RWHYKYNSRI QPIIAVADEG WHILQNKSDD FLLGNHGYDN ALADMHPIFL AHGPAFRKNF SKEAMNSTDL YPLLCHLLNI TAMPHNGSFW NVQDLLNSAM PRVVPYTQST ILLPGSVKPA EYDQEGSYPY FIGVSLGSII VIVFFVIFIK HLIHSQIPAL QDMHAEIAQP LLQA.

This gene encodes a type-I transmembrane glycoprotein. Studies in rat suggest the encoded protein may play a role in neuronal cell communications. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENPP5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1C5-1D7
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ENPP5
Gene Accession No. Q9UJA9
Gene Alias AW061004; D17Abb1e; EC 3.1; ectonucleotide pyrophosphatase/phosphodiesterase 5; ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative); ectonucleotide pyrophosphatase/phosphodiesterase family member 5; E-NPP 5; ENPP5; E-NPP5; KIAA0879; mKIAA4152; Npp5; NPP-5; UNQ550/PRO1107
Gene Symbols ENPP5
Host Species Mouse
Immunogen ENPP5 (AAH27615, 24 a.a. ~ 477 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59084
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.