Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENPP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553515
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553515 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553515 Supplier Invitrogen™ Supplier No. PA553515
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KFWEEATPIW ITNQRAGHTS GAAMWPGTDV KIHKRFPTHY MPYNESVSFE DRVAKIIEWF TSKEPINLGL LYWEDPDDMG HHLGPDSPLM GPVISDIDKK LGYLIQMLKK AKLWNTLNLI ITSDHGMTQC SE Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 86%.

This gene encodes a type-I transmembrane glycoprotein. Studies in rat suggest the encoded protein may play a role in neuronal cell communications. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENPP5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ENPP5
Gene Accession No. Q9UJA9
Gene Alias AW061004; D17Abb1e; EC 3.1; ectonucleotide pyrophosphatase/phosphodiesterase 5; ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative); ectonucleotide pyrophosphatase/phosphodiesterase family member 5; E-NPP 5; ENPP5; E-NPP5; KIAA0879; mKIAA4152; Npp5; NPP-5; UNQ550/PRO1107
Gene Symbols ENPP5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ENPP5. Recombinant protein control fragment (Product #RP-90115).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59084
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.