Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENPP7 Monoclonal Antibody (5E1F9A6)
GREENER_CHOICE

Catalog No. PIMA530371
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530371 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530371 Supplier Invitrogen™ Supplier No. MA530371
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ENPP7/NPP-7.

ENPP7 converts sphingomyelin to ceramide. ENPP7 also has phospholipase C activity toward palmitoyl lyso-phosphocholine. ENPP7 does not appear to have nucleotide pyrophosphatase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENPP7
Applications ELISA
Classification Monoclonal
Clone 5E1F9A6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ENPP7
Gene Accession No. Q6UWV6
Gene Alias alkaline sphingomyelin phosphodiesterase; alkaline sphingomyelinase; Alk-SMase; ectonucleotide pyrophosphatase/phosphodiesterase 7; ectonucleotide pyrophosphatase/phosphodiesterase family member 7; E-NPP 7; ENPP7; E-NPP7; Gm254; Intestinal alkaline sphingomyelinase; NPP7; NPP-7; sphingomyelinase, intestinal alkaline; UNQ3077/PRO9912
Gene Symbols ENPP7
Host Species Mouse
Immunogen Recombinant Human ENPP7 protein (Met1-Ser439).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 339221
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.