Learn More
Invitrogen™ ENPP7 Polyclonal Antibody

Description
ENPP7 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
Specifications
| Antigen | ENPP7 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | ENPP7 |
| Gene Accession No. | Q3TIW9, Q6UWV6 |
| Gene Alias | alkaline sphingomyelin phosphodiesterase; alkaline sphingomyelinase; Alk-SMase; ectonucleotide pyrophosphatase/phosphodiesterase 7; ectonucleotide pyrophosphatase/phosphodiesterase family member 7; E-NPP 7; ENPP7; E-NPP7; Gm254; Intestinal alkaline sphingomyelinase; NPP7; NPP-7; sphingomyelinase, intestinal alkaline; UNQ3077/PRO9912 |
| Gene Symbols | ENPP7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.