Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENPP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102665
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102665 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102665 Supplier Invitrogen™ Supplier No. PA5102665
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ENPP7.

ENPP7 converts sphingomyelin to ceramide. ENPP7 also has phospholipase C activity toward palmitoyl lyso-phosphocholine. ENPP7 does not appear to have nucleotide pyrophosphatase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENPP7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ENPP7
Gene Accession No. Q3TIW9, Q5EZ72, Q6UWV6
Gene Alias alkaline sphingomyelin phosphodiesterase; alkaline sphingomyelinase; Alk-SMase; ectonucleotide pyrophosphatase/phosphodiesterase 7; ectonucleotide pyrophosphatase/phosphodiesterase family member 7; E-NPP 7; ENPP7; E-NPP7; Gm254; Intestinal alkaline sphingomyelinase; NPP7; NPP-7; sphingomyelinase, intestinal alkaline; UNQ3077/PRO9912
Gene Symbols ENPP7
Host Species Rabbit
Immunogen A synthesized peptide derived from human ENPP7(Accession Q6UWV6), corresponding to amino acid residues A174-G224.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 238011, 303729, 339221
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.