Learn More
Description
- Pre-Filled, sterile
- 225 mL buffered peptone water broth, gamma irradiated for immediate use
- Durable Design: Heavyweight, puncture-resistant, multi-laminate polyester pouch ensures secure handling
- Flat Bottom Stability: Stand-up pouch design provides ease during sample addition
- Easy to Seal: Built-in wire tie keeps pouch open for filling and securely closed for transport
- Long Shelf Life: 2 years from date of manufacture for extended storage and readiness
- Certified Quality: ISO 9001:2008 compliant for consistent manufacturing standards Includes
- Filter: Integrated filtration for sample processing
Specifications
Specifications
| Certifications/Compliance | ISO 9001:2008 |
| Color | Clear |
| Media Type | 225 mL buffered peptone water broth |
| Product Type | Enrichment |
| Test Type | Environmental Test |
| For Use With (Application) | Sample preparation and enrichment procedures |
| Includes | No filter |
| Packaging Type | Pouch |
| Quantity | 32/Cs. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
