Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Enrofloxacin Monoclonal Antibody (7F21G1F7E8#1), Invitrogen™
Mouse Monoclonal Antibody
$450.00 - $1108.00
Specifications
Antigen | Enrofloxacin |
---|---|
Clone | 7F21G1F7E8#1 |
Concentration | 1.6 mg/mL |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA546045
|
Invitrogen™
MA546045 |
100 μg |
Each for $450.00
|
|
|||||
01-671-812
|
Invitrogen™
MA110782 |
500 μg |
Each for $602.50
|
|
|||||
PIMA546143
|
Invitrogen™
MA546143 |
1 mg |
Each for $1,108.00
|
|
|||||
Description
MA1-10782 detects Enrofloxacin and its main metabolite, ciproflaxin. MA1-10782 has been successfully used in ELISA applications. The MA1-10782 immunogen is Enrofloxacin conjugated to KLH.
Enrofloxacin is a fluoroquinolone that inhibits the activity of bacterial DNA gyrase. It was the first fluoroquinolone antimicrobial to be used in veterinary medicine and is indicated in therapy of infections by E. coli, Salmonella, Pasteurella, Mycoplasma and Hemophilus species. In the USA, approved use is currently limited to dogs, but approval for use in foodanimals is under consideration.Specifications
Enrofloxacin | |
1.6 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with no preservative; pH 7.4 | |
Enrofloxacin conjugated to KLH. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles |
7F21G1F7E8#1 | |
ELISA | |
Unconjugated | |
Mouse | |
Chemical | |
Baytril | |
IgM | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title