Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enrofloxacin

Catalog No. p-7041721
Click to view available options
Quantity:
5 g
25 g

A plant fungicide and antibacterial agent | CAS: 93106-60-6 | C19H22FN3O3 | 359.40 g/mol

Enrofloxacin is a broad spectrum antibiotic bactericidal agent used in veterinary medicine to treat animals afflicted with certain bacterial infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Enrofloxacin is a broad spectrum antibiotic bactericidal agent used in veterinary medicine to treat animals afflicted with certain bacterial infections.

Solubility
Soluble in chloroform. Slightly soluble in water. Also soluble in dilute KOH

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93106-60-6
Molecular Formula C19H22FN3O3
Molecular Weight (g/mol) 359.40
MDL Number MFCD00792463
InChI Key SPFYMRJSYKOXGV-UHFFFAOYSA-N
Synonym 1-Cyclopropyl-7-(4-ethyl-1-piperazinyl)-6-fluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic acid
PubChem CID 71188
ChEBI CHEBI:35720
IUPAC Name 1-cyclopropyl-7-(4-ethylpiperazin-1-yl)-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
SMILES CCN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1)C(O)=O

Specifications

Type Enrofloxacin
Physical Form Powder
Melting Point 221°C to 226°C
Color White to Yellow
Quantity 5 g
Beilstein 5307824
Merck Index 14,3592
Solubility Information Soluble in chloroform; Slightly soluble in water; Also soluble in dilute KOH
Formula Weight 359.39
Chemical Name or Material Enrofloxacin
RTECSNumber VB1993650
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.