Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENT1 Recombinant Rabbit Monoclonal Antibody (PD00-77)
GREENER_CHOICE

Catalog No. FEMA550728
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550728 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550728 Supplier Invitrogen™ Supplier No. MA550728
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: Human pancreas tissue, human colon carcinoma tissue, human placenta tissue. Predicted band size: 50 kDa Subcellular Location: Basolateral cell membrane, Apical cell membrane, Cell membrane.

This gene is a member of the equilibrative nucleoside transporter family. The gene encodes a transmembrane glycoprotein that localizes to the plasma and mitochondrial membranes and mediates the cellular uptake of nucleosides from the surrounding medium. The protein is categorized as an equilibrative transporter that is sensitive to inhibition by nitrobenzylthioinosine. Nucleoside transporters are required for nucleotide synthesis in cells that lack de novo nucleoside synthesis pathways, and are also necessary for the uptake of cytotoxic nucleosides used for cancer and viral chemotherapies. Multiple alternatively spliced variants, encoding the same protein, have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENT1
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone PD00-77
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SLC29A1
Gene Accession No. Q99808
Gene Alias 1200014D21Rik; AA407560; ENT1; Equilibrative NBMPR-sensitive nucleoside transporter; equilibrative nitrobenzylmercaptopurine riboside (NBMPR)-sensitive nucleoside transporter; equilibrative nitrobenzylmercaptopurine riboside-sensitive nucleoside transporter; Equilibrative nucleoside transporter 1; equilibrative nucleoside transporter 1 variant delta 11; HENT1; NBMPR-sensitive equilibrative nucleoside transporter; nucleoside transporter, es-type; rENT1; SLC29A1; solute carrier family 29 (equilibrative nucleoside transporter), member 1; solute carrier family 29 (nucleoside transporters), member 1; solute carrier family 29 member 1; solute carrier family 29 member 1 (Augustine blood group); solute carrier family 29, member 1
Gene Symbols SLC29A1
Host Species Rabbit
Immunogen Synthetic peptide within C-terminus of human ENT1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2030
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.