Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENT2 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298674MFV500
Description
ENT2 Polyclonal antibody specifically detects ENT2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
ENT2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
Delayed-early response protein 12, DER12Solute carrier family 29 member 2, ENT2, Equilibrative NBMPR-insensitive nucleoside transporter, Equilibrative nitrobenzylmercaptopurine riboside-insensitive nucleosidetransporter, equilibrative nucleoside transporter 2, HNP3636 kDa nucleolar protein HNP36, Hydrophobic nucleolar protein, 36 kDa, hydrophobic nucleolar protein, 36kD, Nucleoside transporter, ei-type, solute carrier family 29 (nucleoside transporters), member 2 | |
Produced in rabbits immunized with E. coli-derived Human ENT2 fragment. | |
0.1 mL | |
Cancer | |
3177 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction