Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ENT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553746
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553746 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553746 Supplier Invitrogen™ Supplier No. PA553746
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KFARYYLANK SSQAQAQELE TKAELLQSDE NGIPSSPQKV ALTLDLDLEK EPESEPDEPQ KPGKPSVFTV FQK Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 70%.

SLC29A2 is a member of the equilibrative nucleoside transporter family which plays a key role in nucleoside and nucleobase uptake for salvage pathways of nucleotide synthesis. SLC29A2 is a transmembrane glycoprotein that mediates the cellular uptake of nucleosides from the surrounding medium. As a nucleoside transporter, SLC29A2 plays an important role in the uptake of nucleoside-based anti-cancer drugs; polymorphisms of point mutations in the gene encoding this protein may affect the efficacy of these drugs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ENT2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC29A2
Gene Accession No. Q14542
Gene Alias 36 kDa hydrophobic nucleolar protein; 36 kDa nucleolar protein HNP36; delayed early response gene 12; delayed-early response protein 12; DER12; ENT2; equilibrative NBMPR-insensitive nucleoside transporter; Equilibrative nitrobenzylmercaptopurine riboside-insensitive nucleoside transporter; equilibrative nucleoside transporter 2; HNP36; hydrophobic nucleolar protein, 36 kDa; hydrophobic nucleolar protein, 36kD; NBMPR-insensitive nucleoside transporter ei; NBMPR-insensitive nucleoside transporter ei 2A; nucleoside transporter, ei-type; rbENT2A; Slc29a2; solute carrier family 29 (equilibrative nucleoside transporter), member 2; solute carrier family 29 (nucleoside transporters), member 2; solute carrier family 29 member 2; solute carrier family 29, member 2
Gene Symbols SLC29A2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC29A2. Recombinant protein control fragment (Product #RP-91298).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3177
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.