Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ENT2 Recombinant Rabbit Monoclonal Antibody (7B2J3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542725
This item is not returnable.
View return policy
Description
Immunogen sequence: MARGDAPRDS YHLVGISFFI LGLGTLLPWN FFITAIPYFQ ARLAGAGNST ARILSTNHTG PEDAFNFNNW VTLLSQLPLL LFTLLNSFLY QCVPETVRIL.
SLC29A2 is a member of the equilibrative nucleoside transporter family which plays a key role in nucleoside and nucleobase uptake for salvage pathways of nucleotide synthesis. SLC29A2 is a transmembrane glycoprotein that mediates the cellular uptake of nucleosides from the surrounding medium. As a nucleoside transporter, SLC29A2 plays an important role in the uptake of nucleoside-based anti-cancer drugs; polymorphisms of point mutations in the gene encoding this protein may affect the efficacy of these drugs.
Specifications
ENT2 | |
Recombinant Monoclonal | |
0.3 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O54699, Q14542, Q61672 | |
SLC29A2 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ENT2/SLC29A2 (Q14542). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
7B2J3 | |
Unconjugated | |
SLC29A2 | |
36 kDa hydrophobic nucleolar protein; 36 kDa nucleolar protein HNP36; delayed early response gene 12; delayed-early response protein 12; DER12; ENT2; equilibrative NBMPR-insensitive nucleoside transporter; Equilibrative nitrobenzylmercaptopurine riboside-insensitive nucleoside transporter; equilibrative nucleoside transporter 2; HNP36; hydrophobic nucleolar protein, 36 kDa; hydrophobic nucleolar protein, 36kD; NBMPR-insensitive nucleoside transporter ei; NBMPR-insensitive nucleoside transporter ei 2A; nucleoside transporter, ei-type; rbENT2A; Slc29a2; solute carrier family 29 (equilibrative nucleoside transporter), member 2; solute carrier family 29 (nucleoside transporters), member 2; solute carrier family 29 member 2; solute carrier family 29, member 2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
13340, 3177, 65194 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction