Learn More
Invitrogen™ Entamoeba histolytica Monoclonal Antibody (3553)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547506
Description
Specificity: Reactive with E. histolytica & E. dispar. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
Entamoeba histolytica is an anaerobic parasitic amoebozoan, part of the genus Entamoeba. Predominantly infecting humans and other primates causing amoebiasis, E. histolytica is estimated to infect about 50 million people worldwide. E. histolytica infection is estimated to kill more than 55,000 people each year.
Specifications
Entamoeba histolytica | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Protozoa | |
Antibody | |
IgG2b |
ELISA, Lateral Flow, Immunocytochemistry | |
3553 | |
Unconjugated | |
E. histolytica | |
Proprietary sequence. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.