Learn More
Invitrogen™ Entamoeba histolytica Monoclonal Antibody (EH147.5)

Description
MA1-10657 detects Entamoeba histolytica. MA1-10657 has been successfully used in ELISA applications. The MA1-10657 immunogen is Entamoeba histolytica HK-9.
Specifications
Specifications
| Antigen | Entamoeba histolytica |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | EH147.5 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | E. histolytica |
| Host Species | Mouse |
| Immunogen | Entamoeba histolytica HK-9. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.