Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Entamoeba histolytica Monoclonal Antibody (EH34.5), Invitrogen™
GREENER_CHOICE

Catalog No. 01671586
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-586 500 μg
PIMA546106 100 μg
PIMA546055 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-671-586 Supplier Invitrogen™ Supplier No. MA110655
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10655 detects Entamoeba histolytica. MA1-10655 has been successfully used in ELISA applications. The MA1-10655 immunogen is Entamoeba histolytica HK-9.

Entamoeba histolytica is an anaerobic parasitic amoebozoan, part of the genus Entamoeba. Predominantly infecting humans and other primates causing amoebiasis, E. histolytica is estimated to infect about 50 million people worldwide. E. histolytica infection is estimated to kill more than 55,000 people each year.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Entamoeba histolytica
Applications ELISA
Classification Monoclonal
Clone EH34.5
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias E. histolytica
Host Species Mouse
Immunogen Entamoeba histolytica HK-9.
Purification Method Protein G
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Protozoa
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.